missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System

Catalog No. 1329871
Change view
Click to view available options
Length (Metric) Cable:
4 m
10 m
Length (English) Cable:
13 ft.
32 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Cable Length (English) Cable
1329871 10 m 32 ft.
1329870 4 m 13 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1329871 Supplier YSI™ Supplier No. 5563 10

Handheld multiprobe meter

DO/Temperature/Conductivity Probe and Cable. Required to measure dissolved oxygen, temperature, conductivity.

Specifications

Length (English) Cable 32 ft.
Length (Metric) Cable 10 m
Includes Temperature/conductivity sensor
Warranty 1 Year
For Use With (Equipment) YSI model 556 handheld multi probe meter
Probe Type DO/Temperature/Conductivity
For Use With (Application) Measures DO, temperature, conductivity