missing translation for 'onlineSavingsMsg'
Learn More

CHECKERS™ Cable Protector Guard Dog

Catalog No. 19085963
Change view
Click to view available options
Includes:
2 channels
5 channels
Diameter (English) Outer:
0.75 in.
1.325 in.
Material:
Metal
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Diameter (English) Outer Material
19085963 2 channels 0.75 in. Metal
19085964 5 channels 1.325 in. Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19085963 Supplier CHECKERS™ Supplier No. GD2X75O/B

Checkers™ Cable Protector Guard Dog is Ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications

  • Color: BLACK
  • Thickness(IN): 0.625
  • Use: Anti-Fatigue

Specifications

Type Dog Bone Connector
Material Metal
For Use With (Equipment) Cables
Color Orange Lid, Black Base
Description Low Profile Cable Protector
Diameter (English) Outer 0.75 in.
Packaging Quantity 1 Ea.
Includes 2 channels