missing translation for 'onlineSavingsMsg'
Learn More

C60MC12 97.0+%, TCI America™

Catalog No. c2415100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2415100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2415100MG Supplier TCI America Supplier No. C2415100MG
Only null left

Chemical Identifiers

CAS 403483-19-2
Molecular Formula C81H35N
Molecular Weight (g/mol) 1022.178
InChI Key YDAHEEMFHYNKKQ-UHFFFAOYSA-N
Synonym C60-fused N-Methylpyrrolidine-m-C12-phenyl
PubChem CID 53384414
SMILES CCCCCCCCCCCCC1=CC=CC(=C1)C2C34C5=C6C7=C8C5=C9C1=C5C%10=C%11C%12=C%13C%10=C%10C1=C8C1=C8C7=C7C%14=C8C8=C%15C(=C%13C8=C%101)C1=C%12C8=C%10C1=C1C%15=C%14C%12=C1C1=C%10C%10=C%13C8=C%11C8=C5C9=C3C(=C%138)C3=C%10C1=C1C%12=C7C6=C1C43CN2C

Specifications

Quantity 100 mg
Formula Weight 1022.18
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material C60MC12