missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ C57BL/6 Mouse Embryonic Fibroblasts, irradiated

Irradiated Gibco C57BL/6 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that support the culture of healthy undifferentiated embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs), especially mouse ESCs/iPSCs.

$122.00

Specifications

Content And Storage Cryopreserved mouse embryonic fibroblasts

Store in liquid nitrogen.
Age Embryonic
Cell Type Feeder Cells (Mouse Embryonic Fibroblast)
Form Cryopreserved
Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. No. of Cells Price Quantity & Availability  
Catalog Number Mfr. No. No. of Cells Price Quantity & Availability  
A34960
Encompass
View Documents Promotion Details
Gibco™
A34960
2 x 106 cells N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A34961
Encompass
View Documents Promotion Details
Gibco™
A34961
4 x 106 cells
Each for $122.00
Only null left
 
Description

Description

Manufactured by MTI-GlobalStem and equivalent to MTI-GlobalStem Cat. Nos. GSC-6002G and GSC-6202G, these MEF cells are isolated from inbred C57BL/6 mice, mitotically arrested by irradiation, and tested to help ensure performance and minimize the risk of contamination.

• Help save precious time with pre-inactivated, ready-to-use MEFs
• Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
• Provide optimum support for mouse ESCs and iPSCs

Help save time with ready-to-use MEFs
Irradiated C57BL/6 MEFs help save time and trouble dealing with an animal facility, performing tedious, time-consuming dissections, and expanding and testing cells day after day, week after week. Focus on your research with these pre-inactivated, ready-to-use feeders.

Culture with meticulously tested feeder cells
Each lot of irradiated C57BL/6 MEFs is tested for bacteria, fungi, mycoplasma, and mouse pathogens to minimize the risk of contamination. Each lot is also tested for post-thaw viability (typically greater than 95%) and ability to support mouse and human ESCs to help ensure robust and consistent performance. Culture your ESCs and iPSCs with confidence, knowing that you can rely on these high quality feeder cells.

Provide optimum support for mouse ESCs and iPSCs
While C57BL/6 MEFs can also support human ESCs and iPSCs, these feeder cells work particularly well for culturing mouse ESCs and iPSCs, showing better results compared to CF1 and ICR strain MEFs.

Specifications

Specifications

Cryopreserved mouse embryonic fibroblasts

Store in liquid nitrogen.
Feeder Cells (Mouse Embryonic Fibroblast)
Mouse
Shipped on dry ice
Embryonic
Cryopreserved
Pooled
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.