Learn More
SOMNI Scientific™ C3 Compact Isoflurane Vaporizer Key Fill
Catalog No. 04355311
Change view
Click to view available options
Animal:
All Animals
Catalog No. Animal
04355311 All Animals
1 options
Catalog No. 04355311 Supplier SOMNI Scientific™ Supplier No. VS6052
The SOMNI C3 Vaporizer is designed with quality materials and built for durability and longevity. The SOMNI C3 is built from brass and specific alloys proven not to react with anesthetic agents. Comes with a stainless steel cagemount.
  • Isoflurane, Key Fill
  • Standard capacity, 125 mL
  • Quality materials
    • Brass, stainless steel
    • Advanced wicking system
  • Calibrated using laser refractometry assures accuracy
  • Safe, leak proof performance
  • One piece fill system
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Animal All Animals
Type Anesthetic Vaporizer