missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ C18-Reversed Phase Silica Gel, Fully Endcapped, 15-25 μm

Catalog No. 111003038
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003038 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003038 Supplier MilliporeSigma™ Supelco™ Supplier No. 60756250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For column chromatography

Reversed-phase silica gels are generally suitable to retain catalysts by hydrophobic interactions.

Specifications

Particle Size 15 to 25 μm
Pore Size 100 Å
Technique Chromatography
Grade For column chromatography
Quantity 250 g
Surface Area (Metric) 380 m2/g
Material Silica
MDL Number MFCD00148338