missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-1-octanol, 95%

Catalog No. p-3593276
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392620500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC392620500 Supplier Thermo Scientific Chemicals Supplier No. 392620500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-21-8 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-21-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00002936
InChI Key PRNCMAKCNVRZFX-UHFFFAOYSA-N
Synonym 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro
PubChem CID 7792
IUPAC Name 3,7-dimethyloctan-1-ol
SMILES CC(C)CCCC(C)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8280g/mL
Boiling Point 118°C (15.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.4360 (20°C, 589nm)
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)2OH
Quantity 50 mL
Specific Gravity 0.828
Formula Weight 158.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 7-Dimethyl-1-octanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 203-374-5

RUO – Research Use Only