missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Basal Cell Culture Liquid Media - Alpha Modification of Eagle's Medium 1X (AMEM)

Catalog No. p-2410140
Change view
Click to view available options
With Additives:
Earle's Balanced Salt Solution (EBSS)
Earle's Balanced Salt Solution (EBSS), Glutamine, Deoxyribonucleosides, Ribonucleosides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. With Additives
MT10022CV Earle's Balanced Salt Solution (EBSS), Glutamine, Deoxyribonucleosides, Ribonucleosides
MT15012CV Earle's Balanced Salt Solution (EBSS)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT10022CV Supplier Corning™ Supplier No. 10022CV

Contains Earle's salts, ribonucleosides, deoxyribonucleosides, and L-glutamine.

  • Format: Liquid

Specifications

Product Type Alpha MEM/EBSS
With Additives Earle's Balanced Salt Solution (EBSS), Glutamine, Deoxyribonucleosides, Ribonucleosides
Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C