missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ C-telopeptide
For cellular and molecular biology applications
Specifications
| Product Type | C-telopeptide |
|---|---|
| Molecular Weight (g/mol) | 868.89 |
| Content And Storage | -30°C to -10°C |
| Sequence | Glu-Lys-Ala-His-Asp-Gly-Gly-Arg |
| Format | Solid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66595EXD
|
Thermo Scientific™
J66595EXD |
0.5 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| C-telopeptide | |
| -30°C to -10°C | |
| Solid | |
| RUO - research use only |
| 868.89 | |
| Glu-Lys-Ala-His-Asp-Gly-Gly-Arg | |
| C-terminal telopeptide | |
| HPLC |
RUO â Research Use Only