missing translation for 'onlineSavingsMsg'
Learn More

Alkanes Mix, C11 through C18 , SPEX CertiPrep™

Catalog No. c1118
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1118 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1118 Supplier SPEX CertiPrep Group LLC Supplier No. C1118
Only null left

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
n-Dodecane112-40-31000μg/mL
n-Heptadecane629-78-71000μg/mL
n-Hexadecane544-76-31000μg/mL
n-Octadecane593-45-31000μg/mL
n-Pentadecane629-62-91000μg/mL
n-Tetradecane629-59-41000μg/mL
n-Tridecane629-50-51000μg/mL
n-Undecane1120-21-41000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material C11 through C18 Alkanes Mix
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Analyte or Component Names Undecane; Dodecane; Tridecane; Tetradecane; Pentadecane; Hexadecane; Heptadecane; Octadecane
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element Undecane, Dodecane, Tridecane, Tetradecane, Pentadecane, Hexadecane, Heptadecane, Octadecane
Solvent or Matrix Methylene Chloride
Industry Type Petrochemical Testing
Quantity 1 mL

Product intended for laboratory use only