Learn More
SignalChem Biotech JAK2, Active 50ug
Catalog No. 502574599
Encompass
Recombinant mouse JAK2 (808-end) was expressed by baculovirus in Sf9 insect cells using a custom N-terminal GST tag and a custom C-terminal tag.
Catalog No. 502574599 Supplier SignalChem Biotech Supplier No. J02M11G50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant mouse JAK2 (808-end) was expressed by baculovirus in Sf9 insect cells using a custom N-terminal GST tag and a custom C-terminal tag.