missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture
GREENER_CHOICE

BVC Fluid Aspiration Systems for Cell Culture are designed for optimal vacuum control, efficient supernatant aspiration, and safe disposal of biological fluids.

Supplier:  BRANDTECH™ VACUUBRAND™ 20727503

 View more versions of this product

Catalog No. 13688014

Only null left

Description

Description

Basic, Control and Professional models to serve your fluid aspiration needs.

  • 2 or 4 L vessels for safe collection and disposal of your biological waste
  • Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
  • The VacuuHandControl (VHC) keeps all fluid within aspiration tubing for the ultimate in containment and cleaning convenience
  • G models feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
  • Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
  • Control and Professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
  • Streamlined design incorporates bottle, controls and pump (Control and Professional models), in a lab friendly small footprint
  • Quick connectors are supplied with the standard (non-G) Professional, but can be added to the other five models

BVC Basic

  • The unpowered version, suitable for use with a VACUU LAN vacuum network, central vacuum system or shared pump
  • Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
  • Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl
  • Available in heavy duty polypropylene, or bleach resistant glass versions

BVC Control

  • The BVC Control features a powerful whisper-quiet VACUUBRAND chemistry-design vacuum pump, eliminating the need for central vacuum supply
  • Potentially biohazardous materials can be contained in a local area
  • The pump is turned on and off automatically on demand when using the VacuuHandControl which further reduces the noise level
  • An easy to clean touch panel adjusts and indicates the vacuum level switch point
  • Available with either a 4 L polypropylene, or 2 L bleach resistant glass bottle

BVC Professional

  • Includes an external liquid level sensor, which operates without contacting bottle contents
  • Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
  • The 4 L polypropylene bottle version is supplied with quick connectors as standard
  • The 2 L glass bottle 'bleach proof' version includes hose barb connections; optional quick connectors are available

Available Accessories:

  • Replacement filter 638266
  • Replacement VHCpro tubing (sold per meter, minimum order 3 m) 636156
  • VHCpro hand controller (one supplied with unit) 688061
  • VHCpro stand 635680
  • Pipette tip ejector for VHCpro 635638
  • 8-channel adapter with tip ejector for VHCpro 635679
  • BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC) 696880
  • Quick connect for second user (between VHCpro and bottle) 635807
  • Standard connect for second user 699943
  • Quick connect (between bottle and pump) 635808
  • Replacement 4 L PP bottle (with cap) 635810
  • Replacement 2 L glass bottle (with cap) 635809
  • 2 L glass bottle adapter 635839
Specifications

Chemical Identifiers

150 mbar (112 Torr)
104°F
10°C
-10°C to +60°C
40°C
1.1 bar

Specifications

150 mbar (112 Torr)
17.9 lb.
2 L Glass Bottle, External Liquid Level Sensor, US Plug, 0.2 μm Biofilter
BVC Professional G
US
16.97 lb.
104°F
10°C
50/60 Hz
43 cm
40.8 cm
Chemistry Diaphragm Pump
1.1 bar
Touchpanel
8.1 kg
100/120 V 50/60 Hz
45 dB (A)
-10°C to +60°C
7.7 kg
40°C
0.4/0.5 CFM
16.92 in.
16.06 in.
7.63 in.
IP40
100/120 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions