missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture

BVC Fluid Aspiration Systems for Cell Culture are designed for optimal vacuum control, efficient supernatant aspiration, and safe disposal of biological fluids.
Supplier: BRANDTECH™ VACUUBRAND™ 20727203
Description
Basic, Control and Professional models to serve your fluid aspiration needs.
- 2 or 4 L vessels for safe collection and disposal of your biological waste
- Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
- The VacuuHandControl (VHC) keeps all fluid within aspiration tubing for the ultimate in containment and cleaning convenience
- G models feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
- Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
- Control and Professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
- Streamlined design incorporates bottle, controls and pump (Control and Professional models), in a lab friendly small footprint
- Quick connectors are supplied with the standard (non-G) Professional, but can be added to the other five models
BVC Basic
- The unpowered version, suitable for use with a VACUU LAN vacuum network, central vacuum system or shared pump
- Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
- Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl
- Available in heavy duty polypropylene, or bleach resistant glass versions
BVC Control
- The BVC Control features a powerful whisper-quiet VACUUBRAND chemistry-design vacuum pump, eliminating the need for central vacuum supply
- Potentially biohazardous materials can be contained in a local area
- The pump is turned on and off automatically on demand when using the VacuuHandControl which further reduces the noise level
- An easy to clean touch panel adjusts and indicates the vacuum level switch point
- Available with either a 4 L polypropylene, or 2 L bleach resistant glass bottle
BVC Professional
- Includes an external liquid level sensor, which operates without contacting bottle contents
- Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
- The 4 L polypropylene bottle version is supplied with quick connectors as standard
- The 2 L glass bottle 'bleach proof' version includes hose barb connections; optional quick connectors are available
Available Accessories:
- Replacement filter 638266
- Replacement VHCpro tubing (sold per meter, minimum order 3 m) 636156
- VHCpro hand controller (one supplied with unit) 688061
- VHCpro stand 635680
- Pipette tip ejector for VHCpro 635638
- 8-channel adapter with tip ejector for VHCpro 635679
- BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC) 696880
- Quick connect for second user (between VHCpro and bottle) 635807
- Standard connect for second user 699943
- Quick connect (between bottle and pump) 635808
- Replacement 4 L PP bottle (with cap) 635810
- Replacement 2 L glass bottle (with cap) 635809
- 2 L glass bottle adapter 635839
Chemical Identifiers
| 150 mbar (112 Torr) | |
| 104°F | |
| 10°C |
| -10°C to +60°C | |
| 40°C | |
| 1.1 bar |
Specifications
| 150 mbar (112 Torr) | |
| 17 lb. | |
| 4 L Polypropylene Bottle, Vacuum Pump, US Plug, 0.2 μm Biofilter | |
| BVC Control | |
| US | |
| 16.09 lb. | |
| 104°F | |
| 10°C | |
| 50/60 Hz | |
| 50 cm | |
| 40.8 cm | |
| Chemistry Diaphragm Pump | |
| 1.1 bar |
| Touchpanel | |
| 7.7 kg | |
| 100/120 V 50/60 Hz | |
| 45 dB (A) | |
| -10°C to +60°C | |
| 7.3 kg | |
| 40°C | |
| 0.4/0.5 CFM | |
| 19.68 in. | |
| 16.06 in. | |
| 7.63 in. | |
| IP40 | |
| 100/120 V |