missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyryl Chloride 98.0+%, TCI America™
$33.56 - $115.57
Chemical Identifiers
| CAS | 141-75-3 |
|---|---|
| Molecular Formula | C4H7ClO |
| Molecular Weight (g/mol) | 106.55 |
| MDL Number | MFCD00000752 |
| InChI Key | DVECBJCOGJRVPX-UHFFFAOYSA-N |
| Synonym | butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride |
| PubChem CID | 8855 |
| IUPAC Name | butanoyl chloride |
| SMILES | CCCC(Cl)=O |
Chemical Identifiers
| 141-75-3 | |
| 106.55 | |
| DVECBJCOGJRVPX-UHFFFAOYSA-N | |
| 8855 | |
| CCCC(Cl)=O |
| C4H7ClO | |
| MFCD00000752 | |
| butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride | |
| butanoyl chloride |
Specifications
| 141-75-3 | |
| 102°C | |
| MFCD00000752 | |
| 2353 | |
| DVECBJCOGJRVPX-UHFFFAOYSA-N | |
| butanoyl chloride | |
| 8855 | |
| ≥98.0% (GC,T) | |
| Butyryl Chloride |
| Yellow | |
| C4H7ClO | |
| 25 g | |
| butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride | |
| CCCC(Cl)=O | |
| 106.55 | |
| 106.55 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes