missing translation for 'onlineSavingsMsg'
Learn More

Butyroin 96.0+%, TCI America™

Catalog No. p-7118774
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09715ML 5 mL
B097125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09715ML Supplier TCI America Supplier No. B09715ML
Only null left

Chemical Identifiers

CAS 496-77-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00021928
InChI Key BVEYJWQCMOVMAR-UHFFFAOYNA-N
Synonym butyroin, 5-hydroxy-4-octanone, 4-octanone, 5-hydroxy, 5-octanol-4-one, octan-4-ol-5-one, fema no. 2587, 5-hydroxy-octan-4-one, acmc-1aijr, ksc236m9n, +/--5-hydroxy-4-octanone
PubChem CID 219794
IUPAC Name 5-hydroxyoctan-4-one
SMILES CCCC(O)C(=O)CCC

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 mL
Formula Weight 144.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyroin
EINECSNumber (2)-0595
TSCA No