missing translation for 'onlineSavingsMsg'
Learn More

Butyroin, 97%

Catalog No. AC460760050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460760050 5 g
AC460760250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460760050 Supplier Thermo Scientific Chemicals Supplier No. 460760050

CAS: 496-77-5 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 496-77-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00021928
InChI Key BVEYJWQCMOVMAR-UHFFFAOYNA-N
Synonym butyroin, 5-hydroxy-4-octanone, 4-octanone, 5-hydroxy, 5-octanol-4-one, octan-4-ol-5-one, fema no. 2587, 5-hydroxy-octan-4-one, acmc-1aijr, ksc236m9n, +/--5-hydroxy-4-octanone
PubChem CID 219794
IUPAC Name 5-hydroxyoctan-4-one
SMILES CCCC(O)C(=O)CCC

Specifications

Boiling Point 80.0°C to 86.0°C (12.0 mmHg)
Flash Point 71°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material Butyroin
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-830-4

RUO – Research Use Only