missing translation for 'onlineSavingsMsg'
Learn More

Butyric Anhydride 98.0+%, TCI America™

Catalog No. p-7118669
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
B076625ML 25 mL
B0766500ML 500 mL
2 options

Catalog No. B076625ML

Supplier: TCI America B076625ML

Only null left

Chemical Identifiers

CAS 106-31-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009389
InChI Key YHASWHZGWUONAO-UHFFFAOYSA-N
Synonym butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
PubChem CID 7798
IUPAC Name butanoyl butanoate
SMILES CCCC(=O)OC(=O)CCC

Specifications

Color Yellow
Boiling Point 201°C
Quantity 25 mL
UN Number 2739
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Butyric Anhydride
EINECSNumber (2)-0627
RTECSNumber ET7090000
TSCA Yes