missing translation for 'onlineSavingsMsg'
Learn More

Butyric Anhydride 98.0+%, TCI America™

Catalog No. B0766500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0766500ML 500 mL
B076625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0766500ML Supplier TCI America Supplier No. B0766500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-31-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009389
InChI Key YHASWHZGWUONAO-UHFFFAOYSA-N
Synonym butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
PubChem CID 7798
IUPAC Name butanoyl butanoate
SMILES CCCC(=O)OC(=O)CCC

Specifications

Color Yellow
Boiling Point 201°C
Quantity 500 mL
UN Number 2739
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Butyric Anhydride
EINECSNumber (2)-0627
RTECSNumber ET7090000
TSCA Yes