Learn More
Butyric anhydride, 98%
Description
Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.
Solubility
Miscible with water and ether.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -75°C |
| Density | 0.968 |
| Boiling Point | 198°C to 199°C |
| Flash Point | 88°C (190°F) |
| Odor | Rancid |
| Refractive Index | 1.413 |
| Linear Formula | (CH3CH2CH2CO)2O |
| Quantity | 2500 mL |
| UN Number | UN2739 |
| Beilstein | 1099474 |
| Show More |
Safety and Handling
| Hazard Category | H227-H314 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYRIC ANHYDRIDE |
| EINECSNumber | 203-383-4 |
| RTECSNumber | ET7090000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only