missing translation for 'onlineSavingsMsg'
Learn More

Butyric anhydride, 98%

Catalog No. AAA12377AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12377AP 500 mL
AAA12377AE 100 mL
AAA123770F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12377AP Supplier Thermo Scientific Chemicals Supplier No. A12377AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-31-0 | C8H14O3 | 158.20 g/mol

Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.

Solubility
Miscible with water and ether.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 106-31-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009389
InChI Key YHASWHZGWUONAO-UHFFFAOYSA-N
Synonym butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
PubChem CID 7798
IUPAC Name butanoyl butanoate
SMILES CCCC(=O)OC(=O)CCC

Specifications

Melting Point -75°C
Density 0.968
Boiling Point 198°C to 199°C
Flash Point 88°C (190°F)
Odor Rancid
Refractive Index 1.413
Linear Formula (CH3CH2CH2CO)2O
Quantity 500 mL
UN Number UN2739
Beilstein 1099474
Sensitivity Moisture sensitive
Merck Index 14,1594
Solubility Information Miscible with water and ether.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Butyric anhydride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYRIC ANHYDRIDE
EINECSNumber 203-383-4
RTECSNumber ET7090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only