missing translation for 'onlineSavingsMsg'
Learn More

Butyric Acid 99.0+%, TCI America™

Catalog No. b075425ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B075425ML 25 mL
B0754500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B075425ML Supplier TCI America Supplier No. B075425ML
Only null left

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

Boiling Point 163°C
Color Colorless
UN Number 2820
Formula Weight 88.11
Percent Purity ≥99.0% (GC,T)
Quantity 25 mL
Physical Form Liquid
Chemical Name or Material Butyric Acid
EINECSNumber (2)-0608
RTECSNumber ES5425000
TSCA Yes