missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyraldoxime 95.0+%, TCI America™
$158.44 - $1575.13
Chemical Identifiers
| CAS | 110-69-0 |
|---|---|
| Molecular Formula | C4H9NO |
| Molecular Weight (g/mol) | 87.122 |
| MDL Number | MFCD00013943 |
| InChI Key | KGGVGTQEGGOZRN-SNAWJCMRSA-N |
| Synonym | butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime |
| PubChem CID | 5324278 |
| IUPAC Name | (NE)-N-butylidenehydroxylamine |
| SMILES | CCCC=NO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B075325ML
|
TCI America
B075325ML |
25 mL |
Each for $158.44
|
|
|||||
|
B0753500ML
|
TCI America
B0753500ML |
500 mL |
Each for $1,575.13
|
|
|||||
Chemical Identifiers
| 110-69-0 | |
| 87.122 | |
| KGGVGTQEGGOZRN-SNAWJCMRSA-N | |
| 5324278 | |
| CCCC=NO |
| C4H9NO | |
| MFCD00013943 | |
| butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime | |
| (NE)-N-butylidenehydroxylamine |
Specifications
| 110-69-0 | |
| 152°C | |
| MFCD00013943 | |
| 2840 | |
| KGGVGTQEGGOZRN-SNAWJCMRSA-N | |
| (NE)-N-butylidenehydroxylamine | |
| 5324278 | |
| ≥95.0% (GC) | |
| Butyraldoxime |
| Yellow | |
| C4H9NO | |
| 25 mL | |
| butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime | |
| CCCC=NO | |
| 87.122 | |
| 87.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0504
RTECSNumber : ES3500000
TSCA : Yes