missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyraldoxime 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 152°C |
| Quantity | 25 mL |
| UN Number | 2840 |
| Formula Weight | 87.12 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Butyraldoxime |
Safety and Handling
| EINECSNumber | (2)-0504 |
| RTECSNumber | ES3500000 |
| TSCA | Yes |