missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde diethylacetal, 97%

Catalog No. AC393430050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393430050 5 g Glass bottle
AC393430250 25 g Glass bottle
AC393431000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC393430050 Supplier Thermo Scientific Chemicals Supplier No. 393430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3658-95-5 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3658-95-5
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00038316
InChI Key UVHXZFGCCJLFMX-UHFFFAOYSA-N
Synonym butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
PubChem CID 77225
IUPAC Name 1,1-diethoxybutane
SMILES CCCC(OCC)OCC

Specifications

Color Colorless
Density 0.8300g/mL
Boiling Point 143.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3950 to 1.3970 (20°C, 589nm)
Linear Formula CH3CH2CH2CH(OCH2CH3)2
Quantity 5 g
Specific Gravity 0.83
Solubility Information Solubility in water: practically insoluble
Formula Weight 146.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Butyraldehyde diethylacetal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-913-5

RUO – Research Use Only