missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde Dibutyl Acetal 98.0+%, TCI America™

Catalog No. p-7123412
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42185G 5 g
B421825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42185G Supplier TCI America Supplier No. B42185G
Only null left

Chemical Identifiers

CAS 5921-80-2
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.338
MDL Number MFCD00561028
InChI Key WQRBJFZKPWPIJD-UHFFFAOYSA-N
Synonym 1,1-Dibutoxybutane
PubChem CID 22210
IUPAC Name 1,1-dibutoxybutane
SMILES CCCCOC(CCC)OCCCC

Specifications

Color Colorless
Boiling Point 214°C
Quantity 5 g
UN Number 1993
Formula Weight 202.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyraldehyde Dibutyl Acetal