Learn More
Butyraldehyde, 99%
CAS: 123-72-8 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals 108090010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Butyraldehyde | |
| -96.0°C | |
| 0.8170g/mL | |
| −12°C | |
| 98.5% min. (GC) | |
| C4H8O | |
| CH3CH2CH2CHO | |
| 1 L | |
| 0.817 | |
| butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal | |
| ZTQSAGDEMFDKMZ-UHFFFAOYSA-N | |
| butanal | |
| 261 | |
| CHEBI:15743 | |
| 99% |
| 123-72-8 | |
| Colorless | |
| 75.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3770 to 1.3830 | |
| MFCD00007023 | |
| 01, 662 | |
| 15, 1595 | |
| Solubility in water: 7.1% (25°C). Other solubilities: miscible with ethanol, ether, ethyl acetate,, acetone and toluene | |
| CCCC=O | |
| 72.11 | |
| 0.43 mPa.s (20°C) | |
| 72.11 | |
| Liquid |
Chemical Identifiers
| 123-72-8 | |
| 72.11 | |
| ZTQSAGDEMFDKMZ-UHFFFAOYSA-N | |
| 261 | |
| butanal |
| C4H8O | |
| MFCD00007023 | |
| butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal | |
| CHEBI:15743 | |
| CCCC=O |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 204-646-6
RUO – Research Use Only