missing translation for 'onlineSavingsMsg'
Learn More

Butylurea 96.0+%, TCI America™

Catalog No. B183125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B183125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B183125G Supplier TCI America Supplier No. B183125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 592-31-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00007956
InChI Key CNWSQCLBDWYLAN-UHFFFAOYSA-N
Synonym n-butylurea, 1-butylurea, urea, butyl, urea, n-butyl, unii-9cpl5nr15k, n-butyl urea, 9cpl5nr15k, dsstox_cid_222, dsstox_rid_75443, dsstox_gsid_20222
PubChem CID 11595
IUPAC Name butylurea
SMILES CCCCNC(=O)N

Specifications

Melting Point 95°C
Quantity 25 g
Formula Weight 116.16
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Butylurea
EINECSNumber (9)-1662
RTECSNumber YS3675000
TSCA Yes