missing translation for 'onlineSavingsMsg'
Learn More

Butyltin Trichloride 98.0+%, TCI America™

Catalog No. B457525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B457525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B457525G Supplier TCI America Supplier No. B457525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1118-46-3
Molecular Formula C4H9Cl3Sn
Molecular Weight (g/mol) 282.176
MDL Number MFCD00000515
InChI Key YMLFYGFCXGNERH-UHFFFAOYSA-K
Synonym butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
PubChem CID 14236
IUPAC Name butyl(trichloro)stannane
SMILES CCCC[Sn](Cl)(Cl)Cl

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 93°C
Quantity 25 g
UN Number 3265
Formula Weight 282.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyltin Trichloride
EINECSNumber (2)-2032&(2)-2285
RTECSNumber WH6780000
TSCA Yes