missing translation for 'onlineSavingsMsg'
Learn More

Butylmalonic Acid 98.0+%, TCI America™

Catalog No. p-7123525
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44145G 5 g
B441425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44145G Supplier TCI America Supplier No. B44145G
Only null left

Chemical Identifiers

CAS 534-59-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00002673
InChI Key MCRZWYDXIGCFKO-UHFFFAOYSA-N
PubChem CID 10801
IUPAC Name 2-butylpropanedioic acid
SMILES CCCCC(C(=O)O)C(=O)O

Specifications

Melting Point 103°C
Color White
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butylmalonic Acid