missing translation for 'onlineSavingsMsg'
Learn More

Butylcyclohexane 99.0+%, TCI America™

Catalog No. p-7118690
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082225ML 25 mL
B0822500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082225ML Supplier TCI America Supplier No. B082225ML
Only null left

Chemical Identifiers

CAS 1678-93-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001530
InChI Key GGBJHURWWWLEQH-UHFFFAOYSA-N
Synonym n-butylcyclohexane, 1-cyclohexylbutane, cyclohexane, butyl, butyl-cyclohexane, butane, 1-cyclohexyl, n-butyl-cyclohexane, 1-cyclohexyl-butane, cyclohexane, n-butyl, acmc-20aj43, butylcyclohexane
PubChem CID 15506
IUPAC Name butylcyclohexane
SMILES CCCCC1CCCCC1

Specifications

Melting Point -78°C
Color Colorless
Boiling Point 180°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butylcyclohexane