missing translation for 'onlineSavingsMsg'
Learn More

Butylchlorodimethylsilane 97.0+%, TCI America™

Catalog No. B201025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B201025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B201025ML Supplier TCI America Supplier No. B201025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1000-50-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00015727
InChI Key MXOSTENCGSDMRE-UHFFFAOYSA-N
Synonym butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
PubChem CID 66079
IUPAC Name butyl(chloro)dimethylsilane
SMILES CCCC[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 2986
Formula Weight 150.72
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butylchlorodimethylsilane
EINECSNumber (2)-2041
TSCA Yes