missing translation for 'onlineSavingsMsg'
Learn More

Butylbenzene 99.0+%, TCI America™

Catalog No. B071325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B071325ML 25 mL
B0713100ML 100 mL
B0713500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B071325ML Supplier TCI America Supplier No. B071325ML

Chemical Identifiers

CAS 104-51-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009463
InChI Key OCKPCBLVNKHBMX-UHFFFAOYSA-N
Synonym n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
PubChem CID 7705
ChEBI CHEBI:44194
IUPAC Name butylbenzene
SMILES CCCCC1=CC=CC=C1

Specifications

Melting Point -88°C
Color Colorless
Boiling Point 183°C
Quantity 25 mL
UN Number 2709
Formula Weight 134.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butylbenzene
EINECSNumber (3)-0011&(3)-0021&(3)-0022
RTECSNumber CY9070000
TSCA Yes