missing translation for 'onlineSavingsMsg'
Learn More

Butylamine 99.0+%, TCI America™

Catalog No. B0707500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0707500ML 500 mL
B070725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0707500ML Supplier TCI America Supplier No. B0707500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-73-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00011690
InChI Key HQABUPZFAYXKJW-UHFFFAOYSA-N
Synonym butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
PubChem CID 8007
ChEBI CHEBI:43799
IUPAC Name butan-1-amine
SMILES CCCCN

Specifications

Melting Point -49°C
Color Colorless
Boiling Point 77°C
Quantity 500 mL
UN Number 1125
Formula Weight 73.14
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Butylamine
EINECSNumber (2)-0132
RTECSNumber EO2975000
TSCA Yes