missing translation for 'onlineSavingsMsg'
Learn More

Butyl Thioglycolate 98.0+%, TCI America™

Catalog No. T090325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T090325ML 25 mL
T0903500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T090325ML Supplier TCI America Supplier No. T090325ML
Only null left

Chemical Identifiers

CAS 10047-28-6
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00022087
InChI Key SKGVGRLWZVRZDC-UHFFFAOYSA-N
Synonym Thioglycolic Acid Butyl Ester, Mercaptoacetic Acid Butyl Ester, Butyl Mercaptoacetate
PubChem CID 82321
IUPAC Name butyl 2-sulfanylacetate
SMILES CCCCOC(=O)CS

Specifications

Color Colorless
Boiling Point 194°C
Quantity 25 mL
UN Number 2810
Formula Weight 148.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Thioglycolate
EINECSNumber (2)-1358
TSCA Yes