missing translation for 'onlineSavingsMsg'
Learn More

Butyl stearate, tech.

Catalog No. AA36338A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36338A3 2 kg
AA3633836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36338A3 Supplier Thermo Scientific Chemicals Supplier No. 036338A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-95-5 | C22H44O2 | 340.59 g/mol

Butyl stearate is used as finishing agents, lubricants and lubricant additives. It is also used as a plasticizer, food packaging material and as a dye solvent. It acts as a reactant and involved in the preparation of octadecanoic acid methyl ester by reacting with methanol. It finds application as a binder in cosmetics like soaps, shampoos and shaving creams, skin conditioners and surfactants for cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-95-5
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.59
MDL Number MFCD00026669
InChI Key ULBTUVJTXULMLP-UHFFFAOYSA-N
Synonym butyl stearate, n-butyl stearate, octadecanoic acid, butyl ester, kesscoflex bs, n-butyl octadecanoate, stearic acid, butyl ester, butyl octadecylate, kessco bsc, stearic acid n-butyl ester, polycizer 332
PubChem CID 31278
ChEBI CHEBI:85983
IUPAC Name butyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCCC

Specifications

Melting Point 17°C to 22°C
Color Colorless to Yellow
Density 0.861 g/mL
Boiling Point 343°C
Flash Point 160°C (320°F)
Refractive Index 1.443
Quantity 2 kg
Beilstein 1792866
Merck Index 14,1589
Solubility Information Immiscible with water. Miscible with ethanol and acetone
Formula Weight 340.59
Grade Technical
Physical Form Liquid
Chemical Name or Material Butyl stearate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-666-5
RTECSNumber WI2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only