missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Stearate 97.0+%, TCI America™
Supplier: TCI America S007725G
Specifications
| Butyl Stearate | |
| White-Yellow | |
| MFCD00026669 | |
| butyl stearate, n-butyl stearate, octadecanoic acid, butyl ester, kesscoflex bs, n-butyl octadecanoate, stearic acid, butyl ester, butyl octadecylate, kessco bsc, stearic acid n-butyl ester, polycizer 332 | |
| CCCCCCCCCCCCCCCCCC(=O)OCCCC | |
| 340.59 | |
| CHEBI:85983 | |
| ≥97.0% (GC) |
| 123-95-5 | |
| C22H44O2 | |
| 25 g | |
| ULBTUVJTXULMLP-UHFFFAOYSA-N | |
| butyl octadecanoate | |
| 31278 | |
| 340.59 | |
| Crystalline Lumps |
Chemical Identifiers
| 123-95-5 | |
| 340.59 | |
| ULBTUVJTXULMLP-UHFFFAOYSA-N | |
| 31278 | |
| butyl octadecanoate |
| C22H44O2 | |
| MFCD00026669 | |
| butyl stearate, n-butyl stearate, octadecanoic acid, butyl ester, kesscoflex bs, n-butyl octadecanoate, stearic acid, butyl ester, butyl octadecylate, kessco bsc, stearic acid n-butyl ester, polycizer 332 | |
| CHEBI:85983 | |
| CCCCCCCCCCCCCCCCCC(=O)OCCCC |
Safety and Handling
EINECSNumber : (2)-0798
RTECSNumber : WI2900000
TSCA : Yes