missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Phthalyl Butyl Glycolate 95.0+%, TCI America™
$52.19 - $309.43
Chemical Identifiers
| CAS | 85-70-1 |
|---|---|
| Molecular Formula | C18H24O6 |
| Molecular Weight (g/mol) | 336.384 |
| MDL Number | MFCD00053793 |
| InChI Key | GOJCZVPJCKEBQV-UHFFFAOYSA-N |
| Synonym | butyl phthalyl butyl glycolate, butoxycarbonylmethyl butyl phthalate, reomol 4pg, butyl carbobutoxymethyl phthalate, santicizer b-16, 2-butoxy-2-oxoethyl butyl phthalate, butyl glycolyl butyl phthalate, butyl phthalate butyl glycolate, caswell no. 131b, dibutyl o-carboxybenzoyloxyacetate |
| PubChem CID | 6819 |
| IUPAC Name | 2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate |
| SMILES | CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC |
Chemical Identifiers
| 85-70-1 | |
| 336.384 | |
| GOJCZVPJCKEBQV-UHFFFAOYSA-N | |
| 6819 | |
| CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC |
| C18H24O6 | |
| MFCD00053793 | |
| butyl phthalyl butyl glycolate, butoxycarbonylmethyl butyl phthalate, reomol 4pg, butyl carbobutoxymethyl phthalate, santicizer b-16, 2-butoxy-2-oxoethyl butyl phthalate, butyl glycolyl butyl phthalate, butyl phthalate butyl glycolate, caswell no. 131b, dibutyl o-carboxybenzoyloxyacetate | |
| 2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate |
Specifications
| 85-70-1 | |
| 345°C | |
| MFCD00053793 | |
| butyl phthalyl butyl glycolate, butoxycarbonylmethyl butyl phthalate, reomol 4pg, butyl carbobutoxymethyl phthalate, santicizer b-16, 2-butoxy-2-oxoethyl butyl phthalate, butyl glycolyl butyl phthalate, butyl phthalate butyl glycolate, caswell no. 131b, dibutyl o-carboxybenzoyloxyacetate | |
| CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC | |
| 336.384 | |
| 336.38 | |
| Liquid |
| Yellow | |
| C18H24O6 | |
| 25 g | |
| GOJCZVPJCKEBQV-UHFFFAOYSA-N | |
| 2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate | |
| 6819 | |
| ≥95.0% (GC) | |
| Butyl Phthalyl Butyl Glycolate |
Safety and Handling
EINECSNumber : (3)-1314
RTECSNumber : TI0535000
TSCA : Yes