missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Palmitate 95.0+%, TCI America™
$298.41 - $298.41
Chemical Identifiers
| CAS | 111-06-8 |
|---|---|
| Molecular Formula | C20H40O2 |
| Molecular Weight (g/mol) | 312.54 |
| MDL Number | MFCD00053738 |
| InChI Key | GLYJVQDYLFAUFC-UHFFFAOYSA-N |
| Synonym | Palmitic Acid Butyl Ester |
| PubChem CID | 8090 |
| IUPAC Name | butyl hexadecanoate |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCCC |
Chemical Identifiers
| 111-06-8 | |
| 312.54 | |
| GLYJVQDYLFAUFC-UHFFFAOYSA-N | |
| 8090 | |
| CCCCCCCCCCCCCCCC(=O)OCCCC |
| C20H40O2 | |
| MFCD00053738 | |
| Palmitic Acid Butyl Ester | |
| butyl hexadecanoate |
Specifications
| 111-06-8 | |
| 170°C | |
| MFCD00053738 | |
| Palmitic Acid Butyl Ester | |
| CCCCCCCCCCCCCCCC(=O)OCCCC | |
| 312.54 | |
| 312.54 | |
| Liquid |
| Yellow | |
| C20H40O2 | |
| 25 mL | |
| GLYJVQDYLFAUFC-UHFFFAOYSA-N | |
| butyl hexadecanoate | |
| 8090 | |
| ≥95.0% (GC) | |
| Butyl Palmitate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes