missing translation for 'onlineSavingsMsg'
Learn More

Butyl p-toluenesulfonate, 99%

Catalog No. p-3777801
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380590250 25 g Glass bottle
AC380591000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC380590250 Supplier Thermo Scientific Chemicals Supplier No. 380590250
Only null left

CAS: 778-28-9 | C11H16O3S | 228.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 778-28-9
Molecular Formula C11H16O3S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00027203
InChI Key QYJXDIUNDMRLAO-UHFFFAOYSA-N
Synonym butyl p-toluenesulfonate, butyl tosylate, n-butyl p-toluenesulfonate, butyl p-methylbenzenesulfonate, butyl-p-toluenesulfonate, p-toluenesulfonic acid, butyl ester, benzenesulfonic acid, 4-methyl-, butyl ester, butyl toluene-4-sulphonate, p-toluenesulfonic acid n-butyl ester, p-toluene sulfonic acid, butyl ester
PubChem CID 13066
IUPAC Name butyl 4-methylbenzenesulfonate
SMILES CCCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Color Brown to Yellow
Density 1.1200g/mL
Boiling Point 170°C to 172°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5030 to 1.505
Linear Formula CH3C6H4SO3(CH2)3CH3
Quantity 25 g
Specific Gravity 1.12
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 228.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyl p-toluenesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-295-5

RUO – Research Use Only