missing translation for 'onlineSavingsMsg'
Learn More

Butyl Oxamate 98.0+%, TCI America™

Catalog No. O021025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O021025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O021025G Supplier TCI America Supplier No. O021025G
Only null left

Chemical Identifiers

CAS 585-28-4
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00059092
InChI Key JQOLMUGUXNNIOR-UHFFFAOYSA-N
Synonym Oxamidic Acid Butyl Ester, Oxamic Acid Butyl Ester
PubChem CID 222466
IUPAC Name butyl 2-amino-2-oxoacetate
SMILES CCCCOC(=O)C(=O)N

Specifications

Melting Point 88°C
Color White
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Butyl Oxamate