missing translation for 'onlineSavingsMsg'
Learn More

Butyl Nitrite 95.0+%, TCI America™

Catalog No. N0376500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0376500ML 500 mL
N037625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0376500ML Supplier TCI America Supplier No. N0376500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002058
InChI Key JQJPBYFTQAANLE-UHFFFAOYSA-N
Synonym 1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
PubChem CID 10996
IUPAC Name butyl nitrite
SMILES CCCCON=O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 500 mL
UN Number 2351
Formula Weight 103.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Nitrite
EINECSNumber (2)-1565
RTECSNumber RA0780000
TSCA Yes
Recommended Storage Refrigerator