missing translation for 'onlineSavingsMsg'
Learn More

Butyl Myristate 97.0+%, TCI America™

Catalog No. M047725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M047725ML 25 mL
M0477500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M047725ML Supplier TCI America Supplier No. M047725ML

Chemical Identifiers

CAS 110-36-1
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00015077
InChI Key DHAZIUXMHRHVMP-UHFFFAOYSA-N
Synonym Myristic Acid Butyl Ester
PubChem CID 8047
IUPAC Name butyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCCCC

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
Formula Weight 284.48
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Myristate
EINECSNumber (2)-0798
RTECSNumber XB8550000
TSCA Yes