missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Methacrylate (stabilized with HQ) 99.0+%, TCI America™
$32.72 - $51.98
Chemical Identifiers
| CAS | 97-88-1 |
|---|---|
| Molecular Formula | C8H14O2 |
| Molecular Weight (g/mol) | 142.20 |
| MDL Number | MFCD00009444 |
| InChI Key | SOGAXMICEFXMKE-UHFFFAOYSA-N |
| Synonym | butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate |
| PubChem CID | 7354 |
| IUPAC Name | butyl 2-methylprop-2-enoate |
| SMILES | CCCCOC(=O)C(C)=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M008125ML
|
TCI America
M008125ML |
25 mL |
Each for $32.72
|
|
|||||
|
M0081500ML
|
TCI America
M0081500ML |
500 mL |
Each for $51.98
|
|
|||||
Chemical Identifiers
| 97-88-1 | |
| 142.20 | |
| SOGAXMICEFXMKE-UHFFFAOYSA-N | |
| 7354 | |
| CCCCOC(=O)C(C)=C |
| C8H14O2 | |
| MFCD00009444 | |
| butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
| butyl 2-methylprop-2-enoate |
Specifications
| 97-88-1 | |
| Colorless | |
| C8H14O2 | |
| 25 mL | |
| butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
| CCCCOC(=O)C(C)=C | |
| 142.20 | |
| 142.20 | |
| Liquid |
| -25°C | |
| 164°C | |
| MFCD00009444 | |
| 2227 | |
| SOGAXMICEFXMKE-UHFFFAOYSA-N | |
| butyl 2-methylprop-2-enoate | |
| 7354 | |
| ≥99.0% (GC) | |
| Butyl Methacrylate (stabilized with HQ) |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ3675000
TSCA : Yes