missing translation for 'onlineSavingsMsg'
Learn More

Butyl Levulinate 98.0+%, TCI America™

Catalog No. p-7131836
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L013825ML 25 mL
L0138500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L013825ML Supplier TCI America Supplier No. L013825ML
Only null left

Chemical Identifiers

CAS 2052-15-5
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00009449
InChI Key ISBWNEKJSSLXOD-UHFFFAOYSA-N
Synonym butyl levulinate, n-butyl levulinate, butyl laevulinate, pentanoic acid, 4-oxo-, butyl ester, n-butyl laevulinate, levulinic acid, butyl ester, butyl 4-ketovalerate, 4-ketopentanoic acid butyl ester, butyl acetylpropionate, n-butyl 4-oxopentanoate
PubChem CID 16331
IUPAC Name butyl 4-oxopentanoate
SMILES CCCCOC(=O)CCC(=O)C

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Levulinate
EINECSNumber (2)-1505
RTECSNumber OI1695000
TSCA Yes