missing translation for 'onlineSavingsMsg'
Learn More

Butyl Laurate 99.0+%, TCI America™

Catalog No. p-7131812
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L008325ML 25 mL
L0083500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L008325ML Supplier TCI America Supplier No. L008325ML
Only null left

Chemical Identifiers

CAS 106-18-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00042870
InChI Key NDKYEUQMPZIGFN-UHFFFAOYSA-N
Synonym butyl laurate, dodecanoic acid, butyl ester, n-butyl laurate, bytyl laurate, butyl dodecylate, lauric acid butyl ester, lauric acid, butyl ester, lauric acid n-butyl ester, unii-23985ocm4h, n-butyl n-dodecanoate
PubChem CID 61015
IUPAC Name butyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCCC

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 mL
Formula Weight 256.43
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Laurate
EINECSNumber (2)-0798
TSCA Yes