missing translation for 'onlineSavingsMsg'
Learn More

Butyl Lactate 98.0+%, TCI America™

Catalog No. L000225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L000225G 25 g
L0002500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L000225G Supplier TCI America Supplier No. L000225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138-22-7
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD00004519
InChI Key MRABAEUHTLLEML-UHFFFAOYNA-N
Synonym butyl lactate, n-butyl lactate, lactic acid, butyl ester, propanoic acid, 2-hydroxy-, butyl ester, butyllactate, 2-hydroxypropanoic acid butyl ester, butylester kyseliny mlecne, butyl alpha-hydroxypropionate, butyl lactate natural, fema no. 2205
PubChem CID 8738
IUPAC Name butyl 2-hydroxypropanoate
SMILES CCCCOC(=O)C(C)O

Specifications

Color Colorless
Boiling Point 187°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Lactate
EINECSNumber (2)-1372&(2)-1373
RTECSNumber OD4025000
TSCA Yes