missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Isovalerate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 176°C |
| Quantity | 25 mL |
| UN Number | 3272 |
| Formula Weight | 158.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Butyl Isovalerate |
Safety and Handling
| EINECSNumber | (2)-0776 |
| RTECSNumber | NY1502000 |
| TSCA | Yes |