missing translation for 'onlineSavingsMsg'
Learn More

Butyl Isovalerate 98.0+%, TCI America™

Catalog No. B371825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B371825ML 25 mL
B3718500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B371825ML Supplier TCI America Supplier No. B371825ML
Only null left

Chemical Identifiers

CAS 109-19-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00048775
InChI Key AYWJSCLAAPJZEF-UHFFFAOYSA-N
Synonym Isovaleric Acid Butyl Ester
PubChem CID 7981
IUPAC Name butyl 3-methylbutanoate
SMILES CCCCOC(=O)CC(C)C

Specifications

Color Colorless
Boiling Point 176°C
Quantity 25 mL
UN Number 3272
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Isovalerate
EINECSNumber (2)-0776
RTECSNumber NY1502000
TSCA Yes