missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Hexanoate 98.0+%, TCI America™
$59.69 - $435.08
Chemical Identifiers
| CAS | 626-82-4 |
|---|---|
| Molecular Formula | C10H20O2 |
| Molecular Weight (g/mol) | 172.268 |
| MDL Number | MFCD00053804 |
| InChI Key | RPRPDTXKGSIXMD-UHFFFAOYSA-N |
| Synonym | Hexanoic Acid Butyl Ester |
| PubChem CID | 12294 |
| IUPAC Name | butyl hexanoate |
| SMILES | CCCCCC(=O)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
H010725ML
|
TCI America
H010725ML |
25 mL |
Each for $59.69
|
|
|||||
|
H0107500ML
|
TCI America
H0107500ML |
500 mL |
Each for $435.08
|
|
|||||
Chemical Identifiers
| 626-82-4 | |
| 172.268 | |
| RPRPDTXKGSIXMD-UHFFFAOYSA-N | |
| 12294 | |
| CCCCCC(=O)OCCCC |
| C10H20O2 | |
| MFCD00053804 | |
| Hexanoic Acid Butyl Ester | |
| butyl hexanoate |
Specifications
| 626-82-4 | |
| 208°C | |
| MFCD00053804 | |
| Hexanoic Acid Butyl Ester | |
| CCCCCC(=O)OCCCC | |
| 172.268 | |
| 172.27 | |
| Liquid |
| Colorless | |
| C10H20O2 | |
| 25 mL | |
| RPRPDTXKGSIXMD-UHFFFAOYSA-N | |
| butyl hexanoate | |
| 12294 | |
| ≥98.0% (GC) | |
| Butyl Hexanoate |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : MO6950000
TSCA : Yes