missing translation for 'onlineSavingsMsg'
Learn More

Butyl Hexanoate 98.0+%, TCI America™

Catalog No. H0107500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0107500ML 500 mL
H010725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0107500ML Supplier TCI America Supplier No. H0107500ML

Chemical Identifiers

CAS 626-82-4
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00053804
InChI Key RPRPDTXKGSIXMD-UHFFFAOYSA-N
Synonym Hexanoic Acid Butyl Ester
PubChem CID 12294
IUPAC Name butyl hexanoate
SMILES CCCCCC(=O)OCCCC

Specifications

Color Colorless
Boiling Point 208°C
Quantity 500 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO6950000
TSCA Yes